5-Iodo-2-(4-morpholino)pyridine 98%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD28798-5GEA
2540
DKK
BLDPBD28798-5G
BLDPBD28798-250MG
BLDPBD28798-100MG
BLDPBD28798-1G
5-Iodo-2-(4-morpholino)pyridine 98%
5-Iodo-2-(4-morpholino)pyridine
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formel:
C₉H₁₁IN₂O Molekylvægt: 290,11 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD04039866 CAS-nummer: 470463-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...