3-Methoxy-4-(trifluoromethyl)benzoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD287022-250MGEA 140 DKK
BLDPBD287022-250MG BLDPBD287022-5G BLDPBD287022-25G BLDPBD287022-1G BLDPBD287022-0.025
3-Methoxy-4-(trifluoromethyl)benzoic acid 95%
3-Methoxy-4-(trifluoromethyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇F₃O₃
Molekylvægt: 220,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13185682
CAS-nummer: 276861-63-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR