3-Amino-6-fluoro-1H-indazole 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD28646-250MGEA 171 DKK
BLDPBD28646-250MG BLDPBD28646-1G BLDPBD28646-10G BLDPBD28646-25G BLDPBD28646-5G
3-Amino-6-fluoro-1H-indazole 97%
3-Amino-6-fluoro-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₆FN₃
Molekylvægt: 151,1400 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06739129
CAS-nummer: 404827-75-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR