2-(3-Pyridinyl)-4-thiazolecarboxylic acid 96%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD28582-250MGEA 225 DKK
BLDPBD28582-250MG BLDPBD28582-5G BLDPBD28582-1G
2-(3-Pyridinyl)-4-thiazolecarboxylic acid 96%
2-(3-Pyridinyl)-4-thiazolecarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₆N₂O₂S
Molekylvægt: 206,22 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00052304
CAS-nummer: 39067-29-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR