Ethyl-2-oxazolecarboxylate 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD28386-10GEA
4670
DKK
BLDPBD28386-10G
BLDPBD28386-1G
BLDPBD28386-250MG
BLDPBD28386-25G
BLDPBD28386-5G
Ethyl-2-oxazolecarboxylate 95%
Ethyl-2-oxazolecarboxylate
Storage condition: Sealed in dry, 2-8°C
Formel:
C₆H₇NO₃ Molekylvægt: 141,1300 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD04114939 CAS-nummer: 33036-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...