5-Bromo-2-chloro-3-hydroxypyridine 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD28266-10GEA
2210
DKK
BLDPBD28266-10G
BLDPBD28266-100MG
BLDPBD28266-25G
BLDPBD28266-100G
BLDPBD28266-250MG
BLDPBD28266-1G
BLDPBD28266-5G
5-Bromo-2-chloro-3-hydroxypyridine 98%
5-Bromo-2-chloro-3-hydroxypyridine
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₅H₃BrClNO Molekylvægt: 208,4400 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08688592 CAS-nummer: 286946-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...