tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate 96%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD28045-5GEA 2690 DKK
BLDPBD28045-5G BLDPBD28045-100MG BLDPBD28045-1G BLDPBD28045-250MG
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate 96%
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₄BrNO₂S
Molekylvægt: 292,193 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03659715
CAS-nummer: 215183-27-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR