[3-(4-Chlorophenyl)-5-isoxazolyl]methanol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27994-250MGEA 714 DKK
BLDPBD27994-250MG BLDPBD27994-100MG BLDPBD27994-1G
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol 97%
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₈ClNO₂
Molekylvægt: 209,6290 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01935957
CAS-nummer: 206055-90-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR