benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27719-1GEA 194 DKK
BLDPBD27719-1G BLDPBD27719-25G BLDPBD27719-5G
benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate 97%
benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₈N₂O₃
Molekylvægt: 262,304 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01568760
CAS-nummer: 167757-45-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR