tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27646-5GEA 54.3 DKK
BLDPBD27646-5G BLDPBD27646-100G BLDPBD27646-25G BLDPBD27646-1G BLDPBD27646-10G
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate 98%
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₂₁NO₃
Molekylvægt: 215,293 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04114966
CAS-nummer: 157634-00-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR