tert-Butyl (4-oxopiperidin-1-yl)acetate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27586-1GEA 3420 DKK
BLDPBD27586-1G BLDPBD27586-250MG BLDPBD27586-100MG
tert-Butyl (4-oxopiperidin-1-yl)acetate 98%
tert-Butyl (4-oxopiperidin-1-yl)acetate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₉NO₃
Molekylvægt: 213,273 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07371646
CAS-nummer: 149554-03-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR