4-Bromo-N-cyclopropyl-2-fluorobenzamide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD275817-250MGEA 256 DKK
BLDPBD275817-250MG BLDPBD275817-1G BLDPBD275817-5G
4-Bromo-N-cyclopropyl-2-fluorobenzamide 98%
4-Bromo-N-cyclopropyl-2-fluorobenzamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₉BrFNO
Molekylvægt: 258,09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09943270
CAS-nummer: 877383-84-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR