6-Nitropyridin-2-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27580-25GEA 15330 DKK
BLDPBD27580-25G BLDPBD27580-1G BLDPBD27580-5G BLDPBD27580-100MG BLDPBD27580-10G BLDPBD27580-250MG
6-Nitropyridin-2-amine 98%
6-Nitropyridin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₅N₃O₂
Molekylvægt: 139,1121 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04114202
CAS-nummer: 14916-63-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR