6-Bromo-1H-pyrrolo[2,3-b]pyridine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27523-25GEA 4550 DKK
BLDPBD27523-25G BLDPBD27523-250MG BLDPBD27523-5G BLDPBD27523-10G BLDPBD27523-1G BLDPBD27523-100G
6-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
6-Bromo-1H-pyrrolo[2,3-b]pyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅BrN₂
Molekylvægt: 197,04 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06659667
CAS-nummer: 143468-13-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR