2,4,6-Triethylaniline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD274570-100MGEA 249 DKK
BLDPBD274570-100MG BLDPBD274570-250MG BLDPBD274570-1G
2,4,6-Triethylaniline 95%
2,4,6-Triethylaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₉N
Molekylvægt: 177,28 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00561929
CAS-nummer: 19779-32-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR