2-(2-Nitrophenyl)-1,3-dioxolane 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273925-1GEA 249 DKK
BLDPBD273925-1G BLDPBD273925-250MG BLDPBD273925-5G
2-(2-Nitrophenyl)-1,3-dioxolane 95%
2-(2-Nitrophenyl)-1,3-dioxolane

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉NO₄
Molekylvægt: 195,172 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06209042
CAS-nummer: 48140-35-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR