3'-Bromo-5'-(trifluoromethyl)acetophenone 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273844-1GEA 574 DKK
BLDPBD273844-1G BLDPBD273844-250MG BLDPBD273844-100MG BLDPBD273844-5G
3'-Bromo-5'-(trifluoromethyl)acetophenone 97%
3'-Bromo-5'-(trifluoromethyl)acetophenone

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆BrF₃O
Molekylvægt: 267,04 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11847339
CAS-nummer: 154259-25-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR