N,N-Diethyl 3-borono-4-methylbenzenesulphonamide 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD273785-1GEA
1370
DKK
BLDPBD273785-1G
BLDPBD273785-250MG
N,N-Diethyl 3-borono-4-methylbenzenesulphonamide 98%
N,N-Diethyl 3-borono-4-methylbenzenesulphonamide
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₁₁H₁₈BNO₄S Molekylvægt: 271,14 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08056356 CAS-nummer: 871329-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...