Thiophene-2,5-diboronic acid bis(pinacol) ester 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273711-1GEA 69.8 DKK
BLDPBD273711-1G BLDPBD273711-25G BLDPBD273711-250MG BLDPBD273711-10G BLDPBD273711-5G BLDPBD273711-100G
Thiophene-2,5-diboronic acid bis(pinacol) ester 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₆H₂₆B₂O₄S
Molekylvægt: 336,06 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD09800563
CAS-nummer: 175361-81-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR