Methyl 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-3-carboxylate 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273651-1GEA 481 DKK
BLDPBD273651-1G BLDPBD273651-100MG BLDPBD273651-5G BLDPBD273651-250MG
Methyl 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-3-carboxylate 96%
Methyl 4'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-3-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₀H₂₃BO₄
Molekylvægt: 338,205 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16294535
CAS-nummer: 1256358-85-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR