[4-(Tetrahydrofurfurylamino-1-carbonyl)phenyl]boronic acid pinacol ester 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273550-250MGEA 256 DKK
BLDPBD273550-250MG BLDPBD273550-5G BLDPBD273550-1G
[4-(Tetrahydrofurfurylamino-1-carbonyl)phenyl]boronic acid pinacol ester 97%
[4-(Tetrahydrofurfurylamino-1-carbonyl)phenyl]boronic acid pinacol ester

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₂₆BNO₄
Molekylvægt: 331,214 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09266200
CAS-nummer: 1073355-09-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR