8-(Trifluoromethyl)quinoline-6-boronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273543-1GEA 4090 DKK
BLDPBD273543-1G BLDPBD273543-100MG BLDPBD273543-250MG
8-(Trifluoromethyl)quinoline-6-boronic acid 98%
8-(Trifluoromethyl)quinoline-6-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₇BF₃NO₂
Molekylvægt: 240,9743 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11504852
CAS-nummer: 1072951-47-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR