2-Amino-2-(3,4-dichlorophenyl)acetic acid hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273447-250MGEA 2800 DKK
BLDPBD273447-250MG BLDPBD273447-100MG
2-Amino-2-(3,4-dichlorophenyl)acetic acid hydrochloride 95%
2-Amino-2-(3,4-dichlorophenyl)acetic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₈Cl₃NO₂
Molekylvægt: 256,51 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115776
CAS-nummer: 1105679-25-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR