Methyl 5-(thiophen-2-yl)isoxazole-3-carboxylate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD273389-1GEA 675 DKK
BLDPBD273389-1G BLDPBD273389-100MG BLDPBD273389-250MG
Methyl 5-(thiophen-2-yl)isoxazole-3-carboxylate 97%
Methyl 5-(thiophen-2-yl)isoxazole-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₇NO₃S
Molekylvægt: 209,22 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03778976
CAS-nummer: 517870-23-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR