Ethyl 5-amino-2-fluorobenzoate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27319-25GEA 2220 DKK
BLDPBD27319-25G BLDPBD27319-1G BLDPBD27319-5G
Ethyl 5-amino-2-fluorobenzoate 98%
Ethyl 5-amino-2-fluorobenzoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₁₀FNO₂
Molekylvægt: 183,18 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09037896
CAS-nummer: 123207-39-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR