1-(3-Methoxyphenyl)ethanamine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD272736-5GEA 543 DKK
BLDPBD272736-5G BLDPBD272736-1G BLDPBD272736-25G BLDPBD272736-250MG
1-(3-Methoxyphenyl)ethanamine 98%
1-(3-Methoxyphenyl)ethanamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₁₃NO
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00673998
CAS-nummer: 62409-13-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR