3-Formyl-N-(tert-butoxycarbonyl)piperidine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27268-100GEA 3330 DKK
BLDPBD27268-100G BLDPBD27268-1G BLDPBD27268-5G
3-Formyl-N-(tert-butoxycarbonyl)piperidine 97%
3-Formyl-N-(tert-butoxycarbonyl)piperidine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₁H₁₉NO₃
Molekylvægt: 213,28 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD02179020
CAS-nummer: 118156-93-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR