3-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD272543-250MGEA 3280 DKK
BLDPBD272543-250MG BLDPBD272543-100MG BLDPBD272543-50MG BLDPBD272543-1G
3-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 95+%
3-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₆BrNO₂
Molekylvægt: 204,02 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD27996611
CAS-nummer: 1639011-56-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR