N-(p-Tolyl)-2-naphthylamine 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD272469-250MGEA 179 DKK
BLDPBD272469-250MG BLDPBD272469-5G BLDPBD272469-1G
N-(p-Tolyl)-2-naphthylamine 98+%
N-(p-Tolyl)-2-naphthylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₇H₁₅N
Molekylvægt: 233,31 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00222912
CAS-nummer: 644-16-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR