6-Methylnaphthalen-1-amine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD272450-250MGEA 1800 DKK
BLDPBD272450-250MG BLDPBD272450-100MG BLDPBD272450-1G
6-Methylnaphthalen-1-amine 95%
6-Methylnaphthalen-1-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₁N
Molekylvægt: 157,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD17012483
CAS-nummer: 116530-22-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR