5-Chloro-3-methylbenzo[c]isoxazole 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD272390-100MGEA 373 DKK
BLDPBD272390-100MG
5-Chloro-3-methylbenzo[c]isoxazole 98%
5-Chloro-3-methylbenzo[c]isoxazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆ClNO
Molekylvægt: 167,59 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD27996551
CAS-nummer: 4104-35-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR