1-Phenyl-1H-pyrazole-4-carboxylic acid 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD27231-1GEA
528
DKK
BLDPBD27231-1G
BLDPBD27231-250MG
BLDPBD27231-5G
1-Phenyl-1H-pyrazole-4-carboxylic acid 95%
1-Phenyl-1H-pyrazole-4-carboxylic acid
Storage condition: Sealed in dry, 2-8°C
Formel:
C₁₀H₈N₂O₂ Molekylvægt: 188,19 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00514518 CAS-nummer: 1134-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...