2,7-Dimethyl-1H-benzo[d]imidazole 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD272277-100MGEA 179 DKK
BLDPBD272277-100MG BLDPBD272277-250MG BLDPBD272277-1G
2,7-Dimethyl-1H-benzo[d]imidazole 95+%
2,7-Dimethyl-1H-benzo[d]imidazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₀N₂
Molekylvægt: 146,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD25372513
CAS-nummer: 23291-66-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR