Methyl 4-amino-3-cyanobenzoate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD272056-10GEA 6650 DKK
BLDPBD272056-10G BLDPBD272056-1G BLDPBD272056-100MG BLDPBD272056-5G BLDPBD272056-250MG
Methyl 4-amino-3-cyanobenzoate 98%
Methyl 4-amino-3-cyanobenzoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₈N₂O₂
Molekylvægt: 176,172 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16038776
CAS-nummer: 159847-80-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR