(S)-Morpholine-3-carboxylic acid hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27175-10GEA 3000 DKK
BLDPBD27175-10G BLDPBD27175-1G BLDPBD27175-5G BLDPBD27175-250MG
(S)-Morpholine-3-carboxylic acid hydrochloride 97%
(S)-Morpholine-3-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₀ClNO₃
Molekylvægt: 167,5900 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06809590
CAS-nummer: 1187929-04-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR