6,7-Dihydroisoquinolin-8(5H)-one hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD271563-1GEA 1160 DKK
BLDPBD271563-1G BLDPBD271563-5G BLDPBD271563-100MG BLDPBD271563-250MG
6,7-Dihydroisoquinolin-8(5H)-one hydrochloride 97%
6,7-Dihydroisoquinolin-8(5H)-one hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₉NO·ClH
Molekylvægt: 183,635 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18432614
CAS-nummer: 135311-97-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR