2-Bromo-5-chloro-4-methylaniline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27128-100MGEA 101 DKK
BLDPBD27128-100MG BLDPBD27128-5G BLDPBD27128-250MG BLDPBD27128-10G BLDPBD27128-1G
2-Bromo-5-chloro-4-methylaniline 95%
2-Bromo-5-chloro-4-methylaniline

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₇BrClN
Molekylvægt: 220,4942 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00806533
CAS-nummer: 102170-52-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR