tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD27077-1GEA 365 DKK
BLDPBD27077-1G BLDPBD27077-250MG BLDPBD27077-25G BLDPBD27077-10G BLDPBD27077-5G
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₁₇N₃O₂S
Molekylvægt: 255,336 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08448157
CAS-nummer: 365996-05-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR