6-Chloro-2-methylpyrido[3,4-d]pyrimidin-4(1H)-one 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD270436-100MGEA 419 DKK
BLDPBD270436-100MG BLDPBD270436-250MG BLDPBD270436-5G BLDPBD270436-1G
6-Chloro-2-methylpyrido[3,4-d]pyrimidin-4(1H)-one 97%
6-Chloro-2-methylpyrido[3,4-d]pyrimidin-4(1H)-one

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₆ClN₃O
Molekylvægt: 195,61 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD27996796
CAS-nummer: 1637671-24-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR