Methyl 3-bromo-4,5-difluorobenzoate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD270327-1GEA 140 DKK
BLDPBD270327-1G BLDPBD270327-5G BLDPBD270327-25G
Methyl 3-bromo-4,5-difluorobenzoate 97%
Methyl 3-bromo-4,5-difluorobenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₅BrF₂O₂
Molekylvægt: 251,025 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20483155
CAS-nummer: 1244642-70-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR