cis-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD270290-100MGEA 163 DKK
BLDPBD270290-100MG BLDPBD270290-250MG
cis-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate 95%
cis-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₁₇FN₂O₂
Molekylvægt: 204,24 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18072719
CAS-nummer: 1431720-86-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR