2-((2,2,2-Trifluoroethyl)amino)ethanol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD270259-1GEA 1280 DKK
BLDPBD270259-1G BLDPBD270259-100MG BLDPBD270259-250MG BLDPBD270259-5G
2-((2,2,2-Trifluoroethyl)amino)ethanol 95%
2-((2,2,2-Trifluoroethyl)amino)ethanol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₄H₈F₃NO
Molekylvægt: 143,10 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD10697861
CAS-nummer: 371-99-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR