Pyrrole-2-carboxylic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD2702-10GEA 179 DKK
BLDPBD2702-10G BLDPBD2702-5G BLDPBD2702-100G BLDPBD2702-25G BLDPBD2702-1G
Pyrrole-2-carboxylic acid 98%
Pyrrole-2-carboxylic acid
Formel: C₅H₅NO₂
Molekylvægt: 111,10 g/mol
MDL Number: MFCD00005219
CAS-nummer: 634-97-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR