2,6-Dimethyl-4-fluorobenzonitrile 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD270091-1GEA 993 DKK
BLDPBD270091-1G BLDPBD270091-250MG BLDPBD270091-5G BLDPBD270091-100MG
2,6-Dimethyl-4-fluorobenzonitrile 98%
2,6-Dimethyl-4-fluorobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₈FN
Molekylvægt: 149,1649 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04972839
CAS-nummer: 14659-61-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR