N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26991-100MGEA 969 DKK
BLDPBD26991-100MG BLDPBD26991-250MG BLDPBD26991-1G
N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide 98+%
N-(4-Chloro-3-iodopyridin-2-yl)-2,2-dimethylpropionamide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₂ClIN₂O
Molekylvægt: 338,58 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08235131
CAS-nummer: 898561-61-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR