Methyl 2-(3-methylpyridin-2-yl)acetate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD269903-1GEA 2010 DKK
BLDPBD269903-1G BLDPBD269903-100MG BLDPBD269903-250MG
Methyl 2-(3-methylpyridin-2-yl)acetate 95%
Methyl 2-(3-methylpyridin-2-yl)acetate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₁NO₂
Molekylvægt: 165,19 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18802818
CAS-nummer: 1092477-78-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR