3-Amino-4-bromo-6-chloropicolinic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD269749-1GEA 5950 DKK
BLDPBD269749-1G BLDPBD269749-100MG BLDPBD269749-50MG BLDPBD269749-250MG
3-Amino-4-bromo-6-chloropicolinic acid 95%
3-Amino-4-bromo-6-chloropicolinic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₄BrClN₂O₂
Molekylvægt: 251,46 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18382761
CAS-nummer: 1073182-90-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR