Ethyl 5-chloro-2-methylbenzoate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD269618-10GEA 3010 DKK
BLDPBD269618-10G BLDPBD269618-250MG BLDPBD269618-1G BLDPBD269618-5G
Ethyl 5-chloro-2-methylbenzoate 97%
Ethyl 5-chloro-2-methylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₁ClO₂
Molekylvægt: 198,64 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11226198
CAS-nummer: 56427-54-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR