7-chloro-benzothiazol-6-ylamine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD269517-100MGEA 373 DKK
BLDPBD269517-100MG BLDPBD269517-1G BLDPBD269517-250MG
7-chloro-benzothiazol-6-ylamine 95%
7-chloro-benzothiazol-6-ylamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₅ClN₂S
Molekylvægt: 184,65 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18822308
CAS-nummer: 70202-00-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR