Methyl-6-bromo-1H-indazole-3-carboxylate 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD26948-5GEA 1320 DKK
BLDPBD26948-5G BLDPBD26948-250MG BLDPBD26948-25G BLDPBD26948-10G BLDPBD26948-1G
Methyl-6-bromo-1H-indazole-3-carboxylate 95%
Methyl-6-bromo-1H-indazole-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₇BrN₂O₂
Molekylvægt: 255,0700 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07371573
CAS-nummer: 885278-42-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR